missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plexin A1 Antibody (708954), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB65361CL1
This item is not returnable.
View return policy
Description
Plexin A1 Monoclonal antibody specifically detects Plexin A1 in Human samples. It is validated for Flow Cytometry,Immunocytochemistry
Specifications
| Plexin A1 | |
| Monoclonal | |
| CoraFluor 1 | |
| NOV, NOVP, plexin A1, PLEXIN-A1 | |
| E. coli-derived recombinant human Plexin A1, Tyr964-Leu1152, Accession # Q9UIW2 | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Immunology, Neuroscience | |
| 5361 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Flow Cytometry, Immunocytochemistry | |
| 708954 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction