missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLET-1 Antibody (700738), mFluor Violet 450 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB6917MFV450
This item is not returnable.
View return policy
Description
PLET-1 Monoclonal antibody specifically detects PLET-1 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF
Specifications
| PLET-1 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 700738 | |
| Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| C11orf34, chromosome 11 open reading frame 34, placenta expressed transcript 1, PLET1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse PLET-1, Ala27-Leu216, Accession # Q8VEN2 | |
| RUO | |
| 349633 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction