missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plerixafor octahydrochloride, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Description
Plerixafor octahydrochloride (AMD3100 octahydrochloride) is a selective CXCR4 antagonist with an IC50 of 44 nM.
Specifications
Specifications
| Chemical Name or Material | Plerixafor octahydrochloride |
| Physical Form | Solid |
| CAS | 155148-31-5 |
| Color | White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C28H62Cl8N8 |
| For Use With (Application) | COVID-19-anti-virus |
| Synonym | AMD3100 octahydrochlorideJM3100 octahydrochlorideSID791 octahydrochloride |
| Solubility Information | H2O : 100 mg/mL (125.87 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction