missing translation for 'onlineSavingsMsg'
Learn More

Plerixafor, MedChemExpress

Product Code. 16424336
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16424336

Brand: Medchem Express HY10046

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Plerixafor (AMD 3100) is a selective CXCR4 antagonist with an IC50 of 44 nM. Plerixafor, an immunostimulant and a hematopoietic stem cell (HSC) mobilizer, is an allosteric agonist of CXCR7. Plerixafor inhibits HIV-1 and HIV-2 replication with an EC50 of 1-10 nM.

Plerixafor (AMD 3100) is a selective CXCR4 antagonist with an IC50 of 44 nM. Plerixafor, an immunostimulant and a hematopoietic stem cell (HSC) mobilizer, is an allosteric agonist of CXCR7. Plerixafor inhibits HIV-1 and HIV-2 replication with an EC50 of 1-10 nM.

Specifications

Chemical Name or Material Plerixafor
Physical Form Solid
CAS 110078-46-1
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C28H54N8
For Use With (Application) COVID-19-anti-virus
Synonym AMD 3100 JM3100 SID791
Solubility Information Ethanol : 50 mg/mL (99.45 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (1.99 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 1 mg/mL (1.99 mM; Need ultrasonic) ∣DMF : 1 mg/mL (1.99 mM; Need ultrasonic)
SMILES C1(CN2CCCNCCNCCCNCC2)=CC=C(C=C1)CN3CCNCCCNCCNCCC3
Molecular Weight (g/mol) 502.78
Formula Weight 502.78
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.