missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ pLenti6.3/V5™-TOPO™ TA Cloning™ Kit

Product Code. 10276743
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10276743 1 kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10276743 Supplier Invitrogen™ Supplier No. K531520

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

pLenti6.3/V5™-TOPO™ TA Cloning™ Kit

Invitrogen's™ pLenti6.3⁄V5™-TOPO™Cloning Kit is part of our ViraPower™ HiPerform™ Lentiviral Expression System (catalog K5310-00).

The pLenti6.3⁄V5™-TOPO™ -TA Cloning Kit contains the TOPO™- adapted ViraPower™ HiPerform™ lentiviral expression vector, pLenti6.3⁄V5™-TOPO™ for quick PCR-based cloning and high-level expression of a target gene in dividing and non-dividing mammalian cells. The pLenti6.3⁄V5™-TOPO™ vector is equipped with two key genetic elements, making it a HiPerform™ vector: the Woodchuck Posttranscriptional Regulatory Element (WPRE) and the central Polypurine Tract (cPPT) sequence from the HIV-1 integrase gene to produce at least 4-fold increase in protein expression compared to vectors lacking these elements.

Advantages
  • Stable expression
  • Long-term experiments
  • Accurate titer of functional virus
  • 5 minute, high-efficiency TOPO™ PCR cloning in three simple steps

    Key Features
  • HiPerform™ WPRE and cPPT elements
  • CMV promoter
  • V5 epitope tag at C terminus
  • Blasticidin selection

    Kit includes
  • A lacZ vector as a positive control, pLenti6.3⁄V5-GW⁄lacZ™
  • Stbl3™ competent cells

    For research use only. Not intended for any therapeutic or diagnostic use.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Cell Type Competent Cell
    Cloning Method TOPO-TA
    Constitutive or Inducible System Constitutive
    Content And Storage TOPO™ TA Cloning Reagents (Box 1)
    pLenti6.3?V5™-TOPO™ (5-10 ng⁄μL): 20 μL, -20°C
    pLenti6.3⁄V5-GW⁄lacZ™ Control vector (0.5 μg⁄μL): 20 μL, -20°C
    10X PCR Buffer: 100 μL, -20°C
    dNTP Mix: 10 μL, -20°C
    Salt Solution: 50 μL, -20°C
    CMV Forward Primer (100 ng⁄μL): 20 μL, -20°C
    V5 (C-term) Reverse Primer (100 ng⁄μL): 20 μL, -20°C
    Control PCR Template (50 ng⁄μL): 10 μL, -20°C
    Control PCR Primers (100 ng⁄μL): 10 μL, each -20°C
    Sterile Water: 1 mL

    One Shot™ Stbl3™ Chemically Competent E. coli (Box 2)
    Stbl3™ Cells: 21 x 50 μL, -80°C
    pUC19 control DNA (10 pg⁄μL): 50 μL, -80°C
    S.O.C. Medium: 6 mL, -80°C
    Delivery Type Lentiviral
    Format Kit
    Expression System ViraPower™ HiPerform™ Lentiviral Expression Systems
    For Use With (Application) Protein Expression
    Key Functions Viral Expression
    Product Line HiPerform, TOPO, ViraPower
    Product Type Cloning Kit
    Promoter CMV
    Protein Tag V5 Epitope Tag
    Quantity 1 kit
    Selection Agent (Eukaryotic) Blasticidin
    Selection Marker Promoter SV40 Promoter
    Vector pLenti, TOPO-TA Vector
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.