missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLEKHO2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559046
This item is not returnable.
View return policy
Description
Immunogen sequence: SVLPDKLKVS WENPSPQEAP AAESAEPSQA PCSETSEAAP REGGKPPTPP PKILSEKLKA SMGEMQASGP PAPGTVQVSV NGMDD Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 59%.
Pleckstrin homology domain-containing proteins family O member 2 (PLEKHO2)'s function remains largely unknown. Genetic polymorphisms of PLEKHO2 contribute to diabetic retinopathy (DR) development. Functional prediction analysis strengthened the likelihood of participation of PLEKHO2 in the development of DR (Eric C. Han et al., 2012).
Specifications
| PLEKHO2 | |
| Polyclonal | |
| Unconjugated | |
| PLEKHO2 | |
| AI840980; PH domain-containing family O member 2; PH domain-containing family Q member 1; PH domain-containing protein; PH domain-containing protein homolog; pleckstrin homology domain containing O2; pleckstrin homology domain containing, family O member 2; pleckstrin homology domain containing, family Q member 1; pleckstrin homology domain-containing family O member 2; Pleckstrin homology domain-containing family Q member 1; PLEKHO2; Plekhq1; PP1628; pp9099 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 80301 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TD55 | |
| PLEKHO2 | |
| Recombinant protein corresponding to Human PLEKHO2. Recombinant protein control fragment (Product #RP-98130). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction