missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLEKHO2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21356-1-AP-20UL
This item is not returnable.
View return policy
Description
Pleckstrin homology domain-containing proteins family O member 2 (PLEKHO2)′s function remains largely unknown. Genetic polymorphisms of PLEKHO2 contribute to diabetic retinopathy (DR) development. Functional prediction analysis strengthened the likelihood of participation of PLEKHO2 in the development of DR (Eric C. Han et al., 2012).Specifications
| PLEKHO2 | |
| Polyclonal | |
| Unconjugated | |
| PLEKHO2 | |
| PLEKHO2, PLEKHQ1, PP1628, pp9099 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 102595, 80301 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8K124, Q8TD55 | |
| Plekho2 | |
| PLEKHO2 Fusion Protein Ag15786 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction