missing translation for 'onlineSavingsMsg'
Learn More

PLEKHO2 Antibody (3D9), Novus Biologicals™

Product Code. 18404719 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18404719 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18404719 Supplier Novus Biologicals Supplier No. H00080301M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PLEKHO2 Monoclonal antibody specifically detects PLEKHO2 in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLEKHO2
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 3D9
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_079477
Gene Alias DKFZp761K2312, FLJ38884, PH domain-containing family O member 2, PH domain-containing family Q member 1, PH domain-containing protein, pleckstrin homology domain containing, family O member 2, pleckstrin homology domain containing, family Q member 1, pleckstrin homology domain-containing family O member 2, Pleckstrin homology domain-containing family Q member 1, PLEKHQ1, PP1628, pp9099
Host Species Mouse
Immunogen pp9099 (NP_079477, 392 a.a. ∽ 490 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. GDLLGEGPRHPLQPRERLYRAQLEVKVASEQTEKLLNKVLGSEPAPVSAETLLSQAVEQLRQATQVLQEMRDLGELSQEAPGLREKRKELVTLYRRSAP
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80301
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.