missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLEKHO1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24883-1-AP-150UL
This item is not returnable.
View return policy
Description
CKIP1 was identified through a yeast-two hybrid screening as a protein that would interact with only one of the two catalytic subunits of the casein kinase 2 complex. CKIP1 is a pleckstrin homology domain-containing protein localized within the nucleus and at the plasma membrane that interacts with CK2alpha but not CK2alpha′, and is thought to play a role in targeting CK2alpha to a particular cellular location. CKIP1 has been implicated in muscle differentiation and the regulation of cell morphology and actin cytoskeleton. CKIP1 can also interact with other proteins such as ATM, an upstream kinase of p53, and recruit the nuclear ATM to the plasma membrane, suggesting CKIP1 may regulate ATM function through re-localizing ATM. CKIP1 can also form a complex with the kinase AKT, leading to a down-regulation of PI3K/AKT signaling and suppression of tumor growth in vivo.Specifications
| PLEKHO1 | |
| Polyclonal | |
| Unconjugated | |
| PLEKHO1 | |
| C Jun binding protein, CK2 interacting protein 1, CKIP 1, CKIP1, CKIP-1, HQ0024c, JBP, OC120, PLEKHO1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51177, 67220 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q53GL0, Q9JIY0 | |
| PLEKHO1 | |
| PLEKHO1 Fusion Protein Ag19309 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction