missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLEKHM3 Polyclonal Antibody

Description
A suggested positive control is mouse skeletal muscle tissue lysate. PA5-20851 can be used with blocking peptide PEP-0965.
PLEKHM3, also known as DAPR, is a member of the M family of Pleckstrin homology domain-containing proteins. PLEKHM3 was initially identified through chromatin immunoprecipitation and CpG microarray analysis examining proteins regulated by myocyte-enhancing factor 2. In C2C12 myoblast cells, PLEKHM3 binds to the PI3K signaling member protein kinase B in the cytosol prior to differentiation into myotubes. Following the initiation of differentiation, PLEKHM3 was also found in membrane fractions. Knockdown of PLEKHM3 expression by RNAi resulted in the inhibition of myotube formation, suggesting that PLEKHM3 is a key component required by myoblasts for orchestrating their differentiation during myogenesis.
Specifications
Specifications
| Antigen | PLEKHM3 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | PLEKHM3 |
| Gene Accession No. | Q6ZWE6, Q8BM47 |
| Gene Alias | 9430067K14Rik; A230102O09Rik; AI449570; Dapr; differentiation associated protein; Differentiation-associated protein; PH domain-containing family M member 3; pleckstrin homology domain containing M3; pleckstrin homology domain containing, family M, member 1-like; pleckstrin homology domain containing, family M, member 3; pleckstrin homology domain-containing family M member 3; PLEKHM1L; Plekhm3; RGD1307151 |
| Gene Symbols | PLEKHM3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?