missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PLEKHM2 Polyclonal Antibody

Product Code. 12838972
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12838972 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12838972 Supplier Invitrogen™ Supplier No. PA520850

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is rat brain tissue lysate. PA5-20850 can be used with blocking peptide PEP-0964.

PLEKHM2, also known as SKIP, is a member of the M family of Pleckstrin homology domain-containing proteins. While little is known of PLEKHM2, a recent study examining differential gene expression in human hematopoietic stem cells has shown it to be specifically expressed in stem cells, suggesting that PLEKHM2 may play a role in erythroid commitment and development. Other studies have shown that PLEKHM2 is required for interaction with the Salmonella virulence factor SifA for Salmonella pathogenesis, suggesting that PLEKHM2 has cellular roles other than in the developing embryo.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLEKHM2
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene PLEKHM2
Gene Accession No. Q80TQ5, Q8IWE5
Gene Alias 2310034J19Rik; AI854247; KIAA0842; mKIAA0842; novel RUN and PH domain-containing protein; PH domain-containing family M member 2; pleckstrin homology and RUN domain containing M2; pleckstrin homology domain containing, family M (with RUN domain) member 2; pleckstrin homology domain-containing family M member 2; Plekhm2; RP11-169K16.1; Salmonella-induced filaments A and kinesin-interacting protein; SifA (Salmonella-induced filaments A) and kinesin-interacting protein; sifA and kinesin-interacting protein; SKIP
Gene Symbols PLEKHM2
Host Species Rabbit
Immunogen A 17 amino acid peptide from near the carboxy terminus of human PLEKHM2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23207, 313667, 69582
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.