missing translation for 'onlineSavingsMsg'
Learn More

PLEKHM2 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18421974 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18421974 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18421974 Supplier Novus Biologicals Supplier No. NBP177089AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PLEKHM2 Polyclonal antibody specifically detects PLEKHM2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLEKHM2
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias KIAA0842RP11-169K16.1, novel RUN and PH domain-containing protein, PH domain-containing family M member 2, pleckstrin homology domain containing, family M (with RUN domain) member 2, pleckstrin homology domain-containing family M member 2, Salmonella-induced filaments A and kinesin-interacting protein, SifA (Salmonella-induced filaments A) and kinesin-interacting protein, SifA and kinesin-interacting protein, SKIP
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide from near the carboxy terminus of human PLEKHM2. The immunogen is located within the last 50 amino acids of PLEKHM2. Amino Acid Squence: TAIQEASNKKKFEDALS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 23207
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.