missing translation for 'onlineSavingsMsg'
Learn More

PLEKHM1 Antibody (1C9), Novus Biologicals™

Product Code. 18358149 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18358149 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18358149 Supplier Novus Biologicals Supplier No. H00009842M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PLEKHM1 Monoclonal antibody specifically detects PLEKHM1 in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLEKHM1
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1C9
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH64361
Gene Alias 162 kDa adapter protein, AP162, hsa-mir-4315-1, KIAA0356, microRNA 4315-1, PH domain-containing family M member 1
Host Species Mouse
Immunogen PLEKHM1 (AAH64361, 957 a.a. ~ 1056 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. PHRFSVADLQQIADGVYEGFLKALIEFASQHVYHCDLCTQRGFICQICQHHDIIFPFEFDTTVRCAECKTVFHQSCQAVVKKGCPRCARRRKYQEQNIFA
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9842
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.