missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLEKHH3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28154-1-AP-20UL
This item is not returnable.
View return policy
Description
28154-1-AP detects the isoforms of PLEKHH3 protein around 80-85 kDa in SDS-PAGE.
PLEKHH3, a gene related to endothelial tight junction, is associated with obstructive sleep apnea (OSA) and angiogenesis. It has been reported that the expression of PLEKHH3 protein was increased in the OSA patients. PLEKHH3 may play a key role in OSA and angiogenesis by specifically regulating the endothelial cells formation of tight junctions.Specifications
| PLEKHH3 | |
| Polyclonal | |
| Unconjugated | |
| PLEKHH3 | |
| FLJ21019, PLEKHH3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 217198, 79990 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q7Z736, Q8VCE9 | |
| PLEKHH3 | |
| PLEKHH3 Fusion Protein Ag28001 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction