missing translation for 'onlineSavingsMsg'
Learn More

PLEKHG5 Antibody [Janelia Fluor™ 549], Novus Biologicals™

Product Code. 30763661 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30763661 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30763661 Supplier Novus Biologicals Supplier No. NBP347117JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PLEKHG5 Polyclonal antibody specifically detects PLEKHG5 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLEKHG5
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias CMTRIC, DSMA4, GEF720, KIAA0720Guanine nucleotide exchange factor 720, NFkB activating protein, novel PH domain-containing protein, PH domain-containing family G member 5, pleckstrin homology domain containing family G member 5, pleckstrin homology domain containing, family G (with RhoGef domain) member 5, pleckstrin homology domain-containing family G member 5, Syx, Tech
Host Species Rabbit
Immunogen pleckstrin homology domain containing, family G(with RhoGef domain) member 5. (Uniprot# O94827)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 57449
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.