missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ PLEKHF2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531268
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
Recommended positive controls: Jurkat, Raji. Predicted reactivity: Mouse (97%), Rat (96%), Zebrafish (82%), Xenopus laevis (82%), Chicken (92%), Rhesus Monkey (99%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PLEKHF2 may play a role in early endosome fusion upstream of RAB5, hence regulating receptor trafficking and fluid-phase transport. Enhances cellular sensitivity to TNF-induced apoptosis.
Tekniske data
| PLEKHF2 | |
| Polyclonal | |
| Unconjugated | |
| PLEKHF2 | |
| 1110070J07Rik; AA673237; EAPF; Endoplasmic reticulum-associated apoptosis-involved protein containing PH and FYVE domains; PH and FYVE domain-containing protein 2; PH domain-containing family F member 2; phafin 2; Phafin2; Phafin-2; pleckstrin homology and FYVE domain containing 2; pleckstrin homology domain containing, family F (with FYVE domain) member 2; pleckstrin homology domain-containing family F member 2; PLEKHF2; ZFYVE18; Zinc finger FYVE domain-containing protein 18 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79666 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.63 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9H8W4 | |
| PLEKHF2 | |
| Recombinant protein encompassing a sequence within the center region of human PLEKHF2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion