missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLEKHF2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | PLEKHF2 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18669270
|
Novus Biologicals
NBP2-93375-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18612721
|
Novus Biologicals
NBP2-93375-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PLEKHF2 Polyclonal antibody specifically detects PLEKHF2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| PLEKHF2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS (pH 7.3), 50% glycerol | |
| 79666 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| FLJ13187, PH and FYVE domain-containing protein 2, Phafin-2, PHAFIN2, pleckstrin homology domain containing, family F (with FYVE domain) member 2, pleckstrin homology domain-containing family F member 2, ZFYVE18PH domain-containing family F member 2, Zinc finger FYVE domain-containing protein 18 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-249 of human PLEKHF2 (NP_078889.1). MVDRLANSEANTRRISIVENCFGAAGQPLTIPGRVLIGEGVLTKLCRKKPKARQFFLFNDILVYGNIVIQKKKYNKQHIIPLENVTIDSIKDEGDLRNGWLIKTPTKSFAVYAATATEKSEWMNHINKCVTDLLSKSGKTPSNEHAAVWVPDSEATVCMRCQKAKFTPVNRRHHCRKCGFVVCGPCSEKRFLLPSQSSKPVRICDFCYDLLSAGDMATCQPARSDSYSQSLKSPLNDMSDDDDDDDSSD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title