missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLEKHB2 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67877-1-IG-150UL
This item is not returnable.
View return policy
Description
The PLEKHB2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PLEKHB2. This antibody recognizes human, pig antigen. The PLEKHB2 antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| PLEKHB2 | |
| Monoclonal | |
| 1000 μg/mL | |
| EVT2; FLJ20783; FLJ23679; FLJ30436 | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human, Pig | |
| 19-25;30-34 kDa | |
| Liquid |
| Western Blot, ELISA | |
| 1H9D7 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 55041 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG1 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur