missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLEKHB2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-68994
This item is not returnable.
View return policy
Description
PLEKHB2 Polyclonal antibody specifically detects PLEKHB2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| PLEKHB2 | |
| Polyclonal | |
| Western Blot 0.04-0.4 μg/ml, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| evectin-2, EVT2FLJ20783, FLJ23679, FLJ30436, PH domain-containing family B member 2, pleckstrin homology domain containing, family B (evectins) member 2, pleckstrin homology domain-containing family B member 2 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: YGYGPYGGAYPPGTQVVYAANGQAYAVPYQYPYAGLYGQQPANQVIIRERYRDNDSDLALGM | |
| 100 μg | |
| Signal Transduction | |
| 55041 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction