missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLEKHA7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584708
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Immunogen sequence: ALKENKDQLE SVLEVLHRQM EQYRDQPQHL EKIAYQQKLL QEDLVHIRAE LSRESTEMEN AWNEYLKLEN DVEQLKQTLQ EQHRRAFFFQ EKSQIQKDLW RIEDVTA Highest antigen sequence indentity to the following orthologs: Mouse - 93%, Rat - 93%.
PLEKHA7 is required for zonula adherens biogenesis and maintenance. Acts via its interaction with KIAA1543/Nezha, which anchors microtubules at their minus-ends to zonula adherens, leading to the recruitment of KIFC3 kinesin to the junctional site.
Spezifikation
| PLEKHA7 | |
| Polyclonal | |
| Unconjugated | |
| PLEKHA7 | |
| A430081P20Rik; AI414265; DKFZp686M22243; Hadp1; Heart adapter protein 1; PH domain-containing family A member 7; PH domain-containing family A member 7 antibody; pleckstrin homology domain containing A7; pleckstrin homology domain containing A7 antibody; pleckstrin homology domain containing, family A member 2; pleckstrin homology domain containing, family A member 7; pleckstrin homology domain-containing family A member 7; Plekha7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 144100 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6IQ23 | |
| PLEKHA7 | |
| Recombinant protein corresponding to Human PLEKHA7. Recombinant protein control fragment (Product #RP-108512). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur