missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLEKHA6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555778
This item is not returnable.
View return policy
Description
Immunogen sequence: SIHEVDISNL EAALRAEEPG GHAYETPREE IARLRKMELE PQHYDVDINK ELSTPDKVLI PERYIDLEPD TPLSPEELKE KQ Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 32%.
PLEKHA6 (Pleckstrin Homology Domain Containing A6) is a Protein Coding gene. Among its related pathways arePI MetabolismandGlycerophospholipid biosynthesis.
Specifications
| PLEKHA6 | |
| Polyclonal | |
| Unconjugated | |
| PLEKHA6 | |
| 9530028L01Rik; AU042621; KIAA0969; LOW QUALITY PROTEIN: pleckstrin homology domain-containing family A member 6; PEPP3; PEPP-3; PH domain-containing family A member 6; phosphoinositol 3-phosphate-binding protein 3; phosphoinositol 3-phosphate-binding protein-3; pleckstrin homology domain containing A6; pleckstrin homology domain containing, family A member 6; pleckstrin homology domain-containing family A member 6; pleckstrin homology domain-containing family A member 6; LOW QUALITY PROTEIN: pleckstrin homology domain-containing family A member 6; Plekha6; RGD1564395 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22874 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y2H5 | |
| PLEKHA6 | |
| Recombinant protein corresponding to Human PLEKHA6. Recombinant protein control fragment (Product #RP-94312). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction