missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLEKHA4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560182
This item is not returnable.
View return policy
Description
Immunogen sequence: PHTEPDSPSP VLQGEESSER ESLPESLELS SPRSPETDWG RPPGGDKDLA SPHLGLGSPR VSRASSPEGR HLPSPQLGTK APVARPRMSA QE Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 76%.
Binds specifically to phosphatidylinositol 3-phosphate (PtdIns3P), but not to other phosphoinositides.
Specifications
| PLEKHA4 | |
| Polyclonal | |
| Unconjugated | |
| PLEKHA4 | |
| PEPP1; PEPP-1; PH domain-containing family A member 4; phosphoinositol 3-phosphate binding protein-1; phosphoinositol 3-phosphate-binding PH domain protein 1; phosphoinositol 3-phosphate-binding protein 1; pleckstrin homology domain containing A4; pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 4; pleckstrin homology domain-containing family A member 4; PLEKHA4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57664 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H4M7 | |
| PLEKHA4 | |
| Recombinant protein corresponding to Human PLEKHA4. Recombinant protein control fragment (Product #RP-98981). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction