missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLEKHA2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06069-100ul
This item is not returnable.
View return policy
Description
PLEKHA2 Polyclonal antibody specifically detects PLEKHA2 in Human samples. It is validated for Western Blot
Specifications
| PLEKHA2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| FLJ25921, PH domain-containing family A member 2, pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 2, pleckstrin homology domain-containing family A member 2, pleckstrin homology domain-containing, family A (phosphoinositide binding specific) member 2, tandem PH Domain containing protein-2, tandem PH domain-containing protein 2, TAPP2, TAPP-2 | |
| Produced in rabbits immunized with E. coli-derived Human PLEKHA2 fragment. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 59339 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction