missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PLEKHA2 Polyclonal antibody specifically detects PLEKHA2 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Gene Alias | FLJ25921, PH domain-containing family A member 2, pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 2, pleckstrin homology domain-containing family A member 2, pleckstrin homology domain-containing, family A (phosphoinositide binding specific) member 2, tandem PH Domain containing protein-2, tandem PH domain-containing protein 2, TAPP2, TAPP-2 |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human PLEKHA2 fragment. |
| Quantity | 0.1 mL |
| Regulatory Status | RUO |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?