missing translation for 'onlineSavingsMsg'
Learn More

PLEKHA2 Antibody, DyLight 594, Novus Biologicals™

Product Code. 18360967 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18360967 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18360967 Supplier Novus Biologicals Supplier No. NBP306069DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PLEKHA2 Polyclonal specifically detects PLEKHA2 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLEKHA2
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 594
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias FLJ25921, PH domain-containing family A member 2, pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 2, pleckstrin homology domain-containing family A member 2, pleckstrin homology domain-containing, family A (phosphoinositide binding specific) member 2, tandem PH Domain containing protein-2, tandem PH domain-containing protein 2, TAPP2, TAPP-2
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human PLEKHA2 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59339
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.