missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLEK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11685-1-AP-20UL
This item is not returnable.
View return policy
Description
Pleckstrin 2 may help orchestrate cytoskeletal arrangement and contribute to lamellipodia formation.Specifications
| PLEK2 | |
| Polyclonal | |
| Unconjugated | |
| PLEK2 | |
| pleckstrin 2, PLEK2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26499, 27260, 314260 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9NYT0, Q9WV52 | |
| PLEK2 | |
| PLEK2 Fusion Protein Ag2297 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction