missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLEK, Mouse anti-Human, Clone: 2D8, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00005341-M03.100ug
This item is not returnable.
View return policy
Description
Sequence: PETIDLGALYLSMKDTEKGIKELNLEKDKKIFNHCFTGNCVIDWLVSNQSVRNRQEGLMIASSLLNEGYLQPAGDMSKSAVDGTAENPFLDNPDAFYYFPDSGFFCEENSSpecifications
| PLEK | |
| Monoclonal | |
| Unconjugated | |
| pleckstrin | |
| FLJ27168/P47 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 2D8 | |
| In 1x PBS, pH 7.4 | |
| BC018549 | |
| PLEK | |
| PLEK (AAH18549, 121 a.a. ∼ 230 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 5341 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction