missing translation for 'onlineSavingsMsg'
Learn More

Pleiotrophin/PTN Antibody (851303R) [mFluor Violet 610 SE], Novus Biologicals™

Product Code. 30732594 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30732594 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732594 Supplier Novus Biologicals Supplier No. FAB2522MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Pleiotrophin/PTN Monoclonal antibody specifically detects Pleiotrophin/PTN in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pleiotrophin/PTN
Applications ELISA
Classification Monoclonal
Clone 851303R
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias HARP, HBBM, HB-GAM, HBGF8, HBNF, HBNF1, HBNF-1, Heparin-binding brain mitogen, NEGF1HBGF-8, neurite growth-promoting factor 1, neurite growth-promoting factor1), OSF-1, Osteoblast-specific factor 1, pleiotrophin
Host Species Rat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Pleiotrophin/PTN, Gly33-Asp168, Accession # P21246
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 5764
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.