missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pleiotrophin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA551122
This item is not returnable.
View return policy
Description
Pleiotrophin Polyclonal Antibody detects endogenous levels of Pleiotrophin protein.
Pleiotrophin Polyclonal antibody specifically detects Pleiotrophin in Human, Mouse, Rat samples. It is validated for ELISA, Western Blot.
Specifications
| Pleiotrophin | |
| Polyclonal | |
| Unconjugated | |
| PTN | |
| HARP; HBBM; HBBN; HB-GAM; HBGF8; HBGF-8; HBNF; HBNF1; HBNF-1; heparin affin regulatory peptide; heparin affin regulatory protein; heparin binding factor; heparin-binding brain mitogen; Heparin-binding growth factor 8; heparin-binding growth-associated molecule; heparin-binding neurite outgrowth promoting factor; Heparin-binding neurite outgrowth-promoting factor; Heparin-binding neurite outgrowth-promoting factor 1; heparin-binding neurite promoting factor; heparin-binding neurotorphic factor; heparin-binding neutrophic factor; NEGF1; OSF; Osf1; OSF-1; osteoblastic cell factor; Osteoblast-specific factor 1; pleiotrophin; pleiotrophin (heparin binding growth factor 8, neurite growth-promoting factor 1); Pleiotrophin (Heparine binding factor, Hbnf, in the mouse); PTN | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 19242, 24924, 5764 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P21246, P63089, P63090 | |
| PTN | |
| Synthesized peptide derived from the Internal region of human PTN. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction