missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Pleiotrophin Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5145351
This item is not returnable.
View return policy
Description
Pleiotrophin (PTN) is an 18 kDa secreted heparin and chondroitin-binding protein that is a regulatory cytokine critical to the early development of blood vessels and neurons. High PTN serum levels are associated with a variety of solid tumors. It was shown that patients with multiple myeloma (MM) also have elevated serum levels of this protein. The expression of this protein is very limited in normal adult tissues.
Specifications
| Pleiotrophin | |
| Polyclonal | |
| Unconjugated | |
| PTN | |
| HARP; HBBM; HBBN; HB-GAM; HBGF8; HBGF-8; HBNF; HBNF1; HBNF-1; heparin affin regulatory peptide; heparin affin regulatory protein; heparin binding factor; heparin-binding brain mitogen; Heparin-binding growth factor 8; heparin-binding growth-associated molecule; heparin-binding neurite outgrowth promoting factor; Heparin-binding neurite outgrowth-promoting factor; Heparin-binding neurite outgrowth-promoting factor 1; heparin-binding neurite promoting factor; heparin-binding neurotorphic factor; heparin-binding neutrophic factor; NEGF1; OSF; Osf1; OSF-1; osteoblastic cell factor; Osteoblast-specific factor 1; pleiotrophin; pleiotrophin (heparin binding growth factor 8, neurite growth-promoting factor 1); Pleiotrophin (Heparine binding factor, Hbnf, in the mouse); PTN | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 19242, 24924, 5764 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| P21246, P63089, P63090 | |
| PTN | |
| Recombinant Human PTN protein, Gly33-Asp168 (Accession #P21246). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction