missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Plectin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115771
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PLEC.
Plectin is a prominent member of an important family of structurally and in part functionally related proteins, termed plakins or cytolinkers, that are capable of interlinking different elements of the cytoskeleton. Plakins, with their multi-domain structure and enormous size, not only play crucial roles in maintaining cell and tissue integrity and orchestrating dynamic changes in cytoarchitecture and cell shape, but also serve as scaffolding platforms for the assembly, positioning, and regulation of signaling complexes. Plectin is expressed as several protein isoforms in a wide range of cell types and tissues from a single gene located on chromosome 8 in humans. The expression of specific plectin isoforms was found to be dependent on cell type (tissue) and stage of development and it appears that each cell type (tissue) contains a unique set (proportion and composition) of plectin isoforms which carry out distinct and specific functions.
Specifications
| Plectin | |
| Polyclonal | |
| Unconjugated | |
| Plec | |
| AA591047; AU042537; EBS1; EBSMD; EBSND; EBSO; EBSOG; EBSPA; HD1; Hemidesmosomal protein 1; LGMD2Q; PCN; Plec; PLEC1; PLEC1b; Plectin; plectin 1; plectin 1, intermediate filament binding protein 500kDa; plectin-1; Plectin-6; Pltn | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 18810, 5339, 64204 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P30427, Q15149, Q9QXS1 | |
| Plec | |
| A synthesized peptide derived from human PLEC(Accession Q15149), corresponding to amino acid residues Q700-R750. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction