missing translation for 'onlineSavingsMsg'
Learn More

Pleconaril, Thermo Scientific™

Product Code. 17525585 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17525585 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17525585 Supplier Thermo Scientific Acros Supplier No. 469221000
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 153168-05-9
Quantity 100 mg
Melting Point 55°C to 65°C
Molecular Formula C18 H18 F3 N3 O3
Formula Weight 381.35
Physical Form Powder
Color White to Beige
Purity ≥97.5%
Chemical Name or Material Pleconaril
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.