missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PLD4 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16683135 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16683135 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16683135 Supplier Invitrogen™ Supplier No. PA5106701

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PLD4.

PLD4 (Phospholipase D Family Member 4) is a Protein Coding gene. Among its related pathways are Innate Immune System and Inositol phosphate metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLD4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PLD4
Gene Accession No. Q8BG07, Q96BZ4
Gene Alias AI132321; C14orf175; Choline phosphatase 4; phosphatidylcholine-hydrolyzing phospholipase D4; phospholipase D family member 4; phospholipase D family, member 4; phospholipase D4; PLD 4; PLD4; thss; UNQ2488/PRO5775
Gene Symbols PLD4
Host Species Rabbit
Immunogen A synthesized peptide derived from human PLD4(Accession Q96BZ4), corresponding to amino acid residues F423-V473.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104759, 122618, 362792
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.