missing translation for 'onlineSavingsMsg'
Learn More

PLD4/Phospholipase D4 Antibody - Azide and BSA Free, Novus Biologicals™

Product Code. 18635930 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.10mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18635930 0.02 mL 0.02mL
18605340 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18635930 Supplier Novus Biologicals Supplier No. NBP2945900.02ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PLD4/Phospholipase D4 Polyclonal antibody specifically detects PLD4/Phospholipase D4 in Human, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLD4/Phospholipase D4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias C14orf175, Choline phosphatase 4, chromosome 14 open reading frame 175, EC 3.1.4.4, Phosphatidylcholine-hydrolyzing phospholipase D4, phospholipase D family, member 4, phospholipase D4, PLD 4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 52-200 of human PLD4 (NP_620145.2). WQVPRPPTWGQVQPKDVPRSWEHGSSPAWEPLEAEARQQRDSCQLVLVESIPQDLPSAAGSPSAQPLGQAWLQLLDTAQESVHVASYYWSLTGPDIGVNDSSSQLGEALLQKLQQLLGRNISLAVATSSPTLARTSTDLQVLAARGAHV
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 122618
Target Species Human, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.