missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLD3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104016
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PLD3.
PLD3 is a ingle-pass type II membrane protein. It belongs to the phospholipase D family. PLD3 contains 2 PLD phosphodiesterase domains. The exact function of PLD3 remains unknown.
Specifications
| PLD3 | |
| Polyclonal | |
| Unconjugated | |
| PLD3 | |
| 5'-3' exonuclease PLD3; phospholipase D3; AD19; Choline phosphatase 3; hindIII K4L homolog; HUK4; HU-K4; phosphatidylcholine-hydrolyzing phospholipase D3; phospholipase D family member 3; phospholipase D family, member 3; phospholipase D3; PLD 3; Pld3; Sam9; Sam-9; Schwannoma-associated protein 9; Unknown (protein for MGC:126910) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18807, 23646, 361527 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O35405, Q5FVH2, Q8IV08 | |
| PLD3 | |
| A synthesized peptide derived from human PLD3(Accession Q8IV08), corresponding to amino acid residues G452-L490. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction