Learn More
Invitrogen™ PLD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587702
Description
Immunogen sequence: MTATPESLFP TGDELDSSQL QMESDEVDTL KEGEDPADRM HPFLAIYELQ SLKVHPLVFA PGVPVTAQVV GTERYTSGSK VGTCTLYSVR LTHGDFSWTT KKKYRHFQEL HRDLLRHKVL MSLLPLARFA VAYSPARDAG NREMPSLPRA GPEGSTRHAA SKQKYLENYL NRLLTMSFYR; Positive Samples: B-cell, HeLa, U-87MG, BT-474, A375; Cellular Location: Membrane, Peripheral membrane protein.
PLD2 is a peripheral membrane protein belonging to phospholipase D family with a PH domain, two PLD phosphodiesterase domains and a PX (phox homology) domain that plays a regulatory rather than catalytic role. PLD2 catalyzes the hydrolysis of phosphatidylcholine (PC) to produce phosphatidic acid and choline. PLD2 may function in regulated secretion, signal-induced cytoskeletal regulation and/or endocytosis, transcriptional regulation, and cell cycle control. It is usually stimulated by phosphatidylinositol 4,5-bisphosphate (PIP2) and activated by the ADP-ribosylation factor-1 (ARF-1). PLD2 is activated by agonist stimulation of both tyrosine kinase and G protein-coupled receptors and is also known to interact with EGFR and PIP5K1A. PLD2, localized to plasma membrane caveolae, is ubiquitously expressed in most tissues.
Specifications
| PLD2 | |
| Polyclonal | |
| Unconjugated | |
| Pld2 | |
| choline phosphatase 2; hPLD2; phosphatidylcholine-hydrolyzing phospholipase D2; phospholipase D gene 2; phospholipase D2; PLD 2; PLD1C; PLD2; Pldc; rPLD2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 18806, 25097 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.55 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P70498, P97813 | |
| Pld2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human PLD2 (NP_0026543). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.