missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£292.00 - £488.00
Specifications
| Antigen | PLD1 |
|---|---|
| Dilution | Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18499941
|
Novus Biologicals
NBP2-34078-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18758893
|
Novus Biologicals
NBP2-34078 |
0.1 mL |
£488.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PLD1 Polyclonal specifically detects PLD1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| PLD1 | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| Q13393 | |
| 5337 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: NEPRVNTSALQKIAADMSNIIENLDTRELHFEGEEVDYDVSPSDPKIQEVYIPFSAIYNTQGFKEPNIQTYLSGCPIKAQVLEVERFTSTTRVPSINLYTIELTHGEFKWQVKRKFKHFQEFHR | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| Choline phosphatase 1, EC 3.1.4.4, hPLD1, Phosphatidylcholine-hydrolyzing phospholipase D1, phospholipase D1, phospholipase D1, phophatidylcholine-specific, phospholipase D1, phosphatidylcholine-specific, PLD 1 | |
| PLD1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title