missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLCXD1 Antibody (OTI2D7), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73458G
This item is not returnable.
View return policy
Description
PLCXD1 Monoclonal specifically detects PLCXD1 in Human samples. It is validated for Immunohistochemistry.
Specifications
| PLCXD1 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PLCXD1 | |
| Full length recombinant protein of human PLCXD1 (NP_060860) produced in E.coli. | |
| 0.1 mL | |
| 55344.0 | |
| Store at 4°C in the dark. |
| Immunohistochemistry | |
| OTI2D7 | |
| Immunohistochemistry | |
| FLJ11323, phosphatidylinositol-specific phospholipase C, X domain containing 1, PI-PLC X domain-containing protein 1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction