Learn More
PLCL2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17421-1-AP-150UL
 View more versions of this product
Product Code. 16857314
  • £349.00 / 150µL
Description
The PLCL2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PLCL2. This antibody recognizes human, mouse, rat antigen. The PLCL2 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Specifications
PLCL2
Polyclonal
Unconjugated
150 μL
23228
IgG
Western Blot, Immunofluorescence, ELISA
267 μg/mL
Rabbit
Primary
Human, Rat, Mouse