missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLCL2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94319-0.02ml
This item is not returnable.
View return policy
Description
PLCL2 Polyclonal antibody specifically detects PLCL2 in Mouse samples. It is validated for Western Blot
Specifications
| PLCL2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| EC 3.1.4.11, FLJ13484, inactive phospholipase C-like protein 2, KIAA1092PLCE2, phospholipase C, epsilon 2, Phospholipase C-epsilon-2, Phospholipase C-L2, phospholipase C-like 2, PLC-epsilon-2, PLC-L(2), PLC-L2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 130-300 of human PLCL2 (NP_055999.2). TLDMLESSQDNMRTSWVSQMFSEIDVDNLGHITLCNAVQCIRNLNPGLKTSKIELKFKELHKSKDKAGTEVTKEEFIEVFHELCTRPEIYFLLVQFSSNKEFLDTKDLMMFLEAEQGVAHINEEISLEIIHKYEPSKEGQEKGWLSIDGFTNYLMSPDCYIFDPEHKKVCQ | |
| 0.02 mL | |
| Signal Transduction | |
| 23228 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction