missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLCG1(phospho Y783) & PLCG1 Protein Phosphorylation Antibody Pair, Abnova™
Antibody Pair
Brand: Abnova DP0166.1Set
This item is not returnable.
View return policy
Description
This protein phosphorylation antibody pair set comes with two antibodies, one against the PLCG1 protein, and the other against the specific Y783 phosphorylated site of PLCG1 for use in in situ Proximity Ligation Assay.Specifications
| PLCG1 | |
| Unconjugated | |
| PLC-II/PLC1/PLC148/PLCgamma1 | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Phospho-PLCG1 Y783 rabbit polyclonal antibody (20 ul) In PBS, 150 mM NaCl, pH 7.4 (0.02% sodium azide, 50% glycerol) 2. PLCG1 mouse monoclonal antibody (40 ug) In 1x PBS, pH 7.2 *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| In situ PLA | |
| phospholipase C, gamma 1 | |
| PLCG1 | |
| RUO | |
| 5335 | |
| Human | |
| Ab Pair |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction