missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLCD4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558264
This item is not returnable.
View return policy
Description
Immunogen sequence: GFTIVFHGRR SNLDLMANSV EEAQIWMRGL QLLVDLVTSM DHQERLDQWL SDWFQRGDKN QDGKMSFQEV QRLLH Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 73%.
This gene encodes a member of the delta class of phospholipase C enzymes. Phospholipase C enzymes play a critical role in many cellular processes by hydrolyzing phosphatidylinositol 4,5-bisphosphate into two intracellular second messengers, inositol 1,4,5-trisphosphate and diacylglycerol. Expression of this gene may be a marker for cancer.
Specifications
| PLCD4 | |
| Polyclonal | |
| Unconjugated | |
| Plcd4 | |
| 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase delta-4; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase delta-4; 4921507K24Rik; hPLCD4; phosphoinositide phospholipase C delta-4; phosphoinositide phospholipase C-delta-4; phospholipase C delta 4; phospholipase C, delta 4; phospholipase C-delta-4; PLC delta4; Plcd; PLCD4; PLC-delta-4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84812 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.9 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BRC7 | |
| Plcd4 | |
| Recombinant protein corresponding to Human PLCD4. Recombinant protein control fragment (Product #RP-102813). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction