missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLCD4 Polyclonal Antibody

Rabbit Polyclonal Antibody
£156.00 - £430.00
Specifications
| Antigen | PLCD4 |
|---|---|
| Concentration | 0.5 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17700754
![]() |
Invitrogen™
PA5119344 |
200 μL |
£430.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17763074
|
Invitrogen™
PA5121678 |
20 μL |
£156.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PLCD4 Polyclonal Antibody for Western Blot, IHC (P)
This gene encodes a member of the delta class of phospholipase C enzymes. Phospholipase C enzymes play a critical role in many cellular processes by hydrolyzing phosphatidylinositol 4,5-bisphosphate into two intracellular second messengers, inositol 1,4,5-trisphosphate and diacylglycerol. Expression of this gene may be a marker for cancer.Specifications
| PLCD4 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse | |
| Plcd4 | |
| 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase delta-4; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase delta-4; 4921507K24Rik; hPLCD4; phosphoinositide phospholipase C delta-4; phosphoinositide phospholipase C-delta-4; phospholipase C delta 4; phospholipase C, delta 4; phospholipase C-delta-4; PLC delta4; Plcd; PLCD4; PLC-delta-4 | |
| Plcd4 | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q8K3R3, Q9BRC7 | |
| 18802, 84812 | |
| Recombinant protein Phospholipase C Delta 4. The antigen corresponds to amino acid range 1-250 of the target protein. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title