missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLCD4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94312-0.1ml
This item is not returnable.
View return policy
Description
PLCD4 Polyclonal antibody specifically detects PLCD4 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| PLCD4 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| EC 3.1.4.11, hPLCD4, MGC12837, Phosphoinositide phospholipase C-delta-4, phospholipase C, delta 4, Phospholipase C-delta-4, PLC delta4, PLC-delta-4,1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase delta-4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-290 of human PLCD4 (NP_116115.1). MASLLQDQLTTDQDLLLMQEGMPMRKVRSKSWKKLRYFRLQNDGMTVWHARQARGSAKPSFSISDVETIRNGHDSELLRSLAEELPLEQGFTIVFHGRRSNLDLMANSVEEAQIWMRGLQLLVDLVTSMDHQERLDQWLSDWFQRGDKNQDGKMSFQEVQRLLHLMNVEMDQEYAFSLFQAADTSQSGTLEGEEFVQFYKALTKRAEVQELFESFSADGQKLTLLEFLDFLQEEQKERDCTSELALELIDRYEPSDSGKLRHVLSMDGFLSYLCSKDGDIFNPACLPIYQ | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 84812 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction