missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLCD3 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547717
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
This gene encodes a member of the phospholipase C family, which catalyze the hydrolysis of phosphatidylinositol 4, 5-bisphosphate to generate the second messengers diacylglycerol and inositol 1, 4, 5-trisphosphate (IP3). Diacylglycerol and IP3 mediate a variety of cellular responses to extracellular stimuli by inducing protein kinase C and increasing cytosolic Ca(2+) concentrations. This enzyme localizes to the plasma membrane and requires calcium for activation. Its activity is inhibited by spermine, sphingosine, and several phospholipids.
Specifications
| PLCD3 | |
| Polyclonal | |
| Unconjugated | |
| PLCD3 | |
| 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase delta-3; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase delta-3; 2610205J15Rik; Kiaa1964; mKIAA1964; phosphoinositide phospholipase C-delta-3; phospholipase C delta 3; phospholipase C, delta 3; phospholipase C-delta-3; PLC delta3; Plcd3; PLC-delta-3 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 113026, 72469 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q8K2J0, Q8N3E9 | |
| PLCD3 | |
| E. coli-derived recombinant human PLC-delta3 Lys148-Ala344. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction