Learn More
Invitrogen™ PLCB3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120077
Description
Positive test controls include: OVCAR8, Mouse lung, Rat lung. The target is usually found in the following locations: Membrane, Peripheral membrane protein. Immunogen sequence: GALGGAADVE DTKEGEDEAK RYQEFQNRQV QSLLELREAQ VDAEAQRRLE HLRQALQRLR EVVLDANTTQ FKRLKEMNER EKKELQKILD RKRHNSISEA KMRDKHKKEA ELTEINRRHI TESVNSIRRL EEAQKQRHDR LVAGQQQVLQ QLAEEEPKLL AQLAQECQEQ RARLPQEIRR SLLGEMPEGL GDGPLVACAS NGHAPGSSGH LSGADSESQE ENTQL.
PLCB3 encodes a member of the phosphoinositide phospholipase C beta enzyme family that catalyze the production of the secondary messengers diacylglycerol and inositol 1,4,5-triphosphate from phosphatidylinositol in G-protein-linked receptor-mediated signal transduction. Alternative splicing results in multiple transcript variants.
Specifications
| PLCB3 | |
| Polyclonal | |
| Unconjugated | |
| Plcb3 | |
| 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase beta-3; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta-3; EC 3.1.4.11; mKIAA4098; Phosphoinositide phospholipase C-beta-3; phosphoinositide-specific phospholipase c; phospholipase C beta 3; phospholipase C, beta 3; phospholipase C, beta 3 (phosphatidylinositol-specific); phospholipase C-beta-3; PLC beta 3; Plcb3; PLCbeta3; PLC-beta3; PLC-beta-3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 18797, 29322, 5331 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.65 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P51432, Q01970, Q99JE6 | |
| Plcb3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1010-1234 of human PLC beta 3 (PLCB3) (NP_001303243.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.