Learn More
Invitrogen™ PLCB2 Monoclonal Antibody (K94011_8D9), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555708
Description
Sequence of this protein is as follows: EGPEGVDGRV RELKDRLELE LLRQGEEQYE CVLKRKEQHV AEQISKMMEL AREKQAAELK ALKETSENDT KEMKKKLETK RLERIQGMTK VTTDKMAQER LKREINNSHI QEVVQVIKQM TENLERHQEK LEEKQAACLE QIREMEKQFQ KEALAEYEAR MKGLEAEVKE SVRACLRTCF PSEAKDKPER ACECPPELCE QDPLIAKADA
PLCB2 is the production of the second messenger molecules diacylglycerol (DAG) and inositol 1,4,5-trisphosphate (IP3) is mediated by activated phosphatidylinositol-specific phospholipase C enzymes.
Specifications
| PLCB2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q00722 | |
| PLCB2 | |
| Recombinant human PLCB2 (immunogen range is 971-1180). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| K94011_8D9 | |
| Unconjugated | |
| PLCB2 | |
| 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase beta-2; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta-2; AI550384; B230205M18Rik; B230399N12; Phosphoinositide phospholipase C-beta-2; phospholipase C beta 2; phospholipase C, beta 2; phospholipase C-beta-2; PKC beta II; PLC beta-2; PLCB2; plcb2 {ECO:0000250; PLCbeta2; PLC-beta2; PLC-beta-2; PLCbeta-2; UniProtKB:Q00722} | |
| Mouse | |
| Protein A | |
| RUO | |
| 5330 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.