missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLCB2 Monoclonal Antibody (K94011_16H7), Invitrogen™
Mouse Monoclonal Antibody
£380.00 - £570.00
Specifications
| Antigen | PLCB2 |
|---|---|
| Clone | K94011_16H7 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30195950
|
Invitrogen™
MA555711 |
50 μL |
£380.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30204686
|
Invitrogen™
MA556091 |
200 μL |
£570.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: EGPEGVDGRV RELKDRLELE LLRQGEEQYE CVLKRKEQHV AEQISKMMEL AREKQAAELK ALKETSENDT KEMKKKLETK RLERIQGMTK VTTDKMAQER LKREINNSHI QEVVQVIKQM TENLERHQEK LEEKQAACLE QIREMEKQFQ KEALAEYEAR MKGLEAEVKE SVRACLRTCF PSEAKDKPER ACECPPELCE QDPLIAKADA
PLCB2 Monoclonal antibody specifically detects PLCB2 in Human samples. It is validated for Western BlotSpecifications
| PLCB2 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase beta-2; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta-2; AI550384; B230205M18Rik; B230399N12; Phosphoinositide phospholipase C-beta-2; phospholipase C beta 2; phospholipase C, beta 2; phospholipase C-beta-2; PKC beta II; PLC beta-2; PLCB2; plcb2 {ECO:0000250; PLCbeta2; PLC-beta2; PLC-beta-2; PLCbeta-2; UniProtKB:Q00722} | |
| Recombinant human PLCB2 (immunogen range is 971-1180). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| PLCB2 |
| K94011_16H7 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q00722 | |
| 5330 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title