missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLCB1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119929
This item is not returnable.
View return policy
Description
Positive Control: Rat brain tissue lysate, human brain tissue lysate, mouse brain tissue lysates, rat brain tissue, mouse brain tissue, SH-SY5Y. Subcellular Location: Nucleus membrane, Cytoplasm.
The protein encoded by this gene catalyzes the formation of inositol 1,4,5-trisphosphate and diacylglycerol from phosphatidylinositol 4,5-bisphosphate. This reaction uses calcium as a cofactor and plays an important role in the intracellular transduction of many extracellular signals. This gene is activated by two G-protein alpha subunits, alpha-q and alpha-11.
Specifications
| PLCB1 | |
| Polyclonal | |
| Unconjugated | |
| PLCB1 | |
| 1-phosphatidyl-D-myo-inositol-4,5-bisphosphate; 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase beta-1; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta-1; 3110043I21Rik; AI132408; EIEE12; inositoltrisphosphohydrolase; KIAA0581; mKIAA0581; monophosphatidylinositol phosphodiesterase; Phosphb; phosphoinositidase C; phosphoinositide phospholipase C; phosphoinositide phospholipase C-beta-1; phosphoinositide-specific phospholipase c; phospholipase C; phospholipase C beta 1; phospholipase C, beta 1; phospholipase C, beta 1 (phosphoinositide-specific); Phospholipase C-beta1; phospholipase C-beta-1; phospholipase C-beta-1a; phospholipase C-I; PI-PLC; PLC'1; PLC154; PLC-154; Plcb; PLCB1; PLCB1A; PLCB1B; PLCbeta1; PLC-beta-1; PLC-I; RATPHOSPHB; triphosphoinositide phosphodiesterase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18795, 23236, 24654 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| P10687, Q9NQ66, Q9Z1B3 | |
| PLCB1 | |
| Recombinant protein within human Phospholipase C beta 1 aa 50-250/1216. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction