missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLC-gamma 2 Antibody (3A8B6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£173.00 - £416.00
Specifications
| Antigen | PLC-gamma 2 |
|---|---|
| Clone | 3A8B6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18261030
|
Novus Biologicals
NBP2-52534 |
0.1 mg |
£416.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18631388
|
Novus Biologicals
NBP2-52534-0.025mg |
0.025 mg |
£173.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PLC-gamma 2 Monoclonal specifically detects PLC-gamma 2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, CyTOF-ready.Specifications
| PLC-gamma 2 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, CyTOF | |
| Unconjugated | |
| Mouse | |
| Lipid and Metabolism, Phospho Specific | |
| PBS with 0.05% Sodium Azide | |
| 5336 | |
| Synthesized peptide of human PLCG2 (AA: phospho-Tyrosine 753 of human Phospholipase Cg2(cERDINSLpYDVSRMYV)). | |
| Primary | |
| 148 kDa |
| 3A8B6 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 3.1.4.11, Phosphoinositide phospholipase C-gamma-2, phospholipase C gamma 2, phospholipase C, gamma 2 (phosphatidylinositol-specific), phospholipase C, gamma 2 (phosphatidylyinositol-specific), Phospholipase C-gamma-2,1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase gamma-2, Phospholipase C-IV, PLCG2, PLC-gamma-2, PLC-IV | |
| PLCG2 | |
| IgG2b | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title